Avantor® Alltima HP, HILIC HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica. The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
- Excellent column stability
- Low to no detectable column bleed
- Multiple selectivity options
Hydrophilic interaction chromatography uses small amounts of water for increased sensitivity with microbore applications.
Very polar analytes that cannot be retained by reversed-phase.
USP code L3.
Customers Who Bought This Also Bought