A propos de cet article
SterilEnz®-II/EC Polypropylene End Caps are the only End Caps that come equipped with a medical grade silicone gasket pre-attached to the fitting face. This feature prevents operators from misaligning or dropping gaskets during assembly.
- Standard sanitary clamp fitting per ASME BPE specifications
- Intended for single-use applications
- Built-in gasket eliminates gasket preparation
- Easy hold tab for easy on/off installation
- Gaskets cannot be misaligned
- No leaks or potential contamination issues
- Certified free from any animal derived components or processes
- Animal-free
- Gamma-stable, for use in irradiated disposable systems, 50kGy
- Autoclaveable at 123°C for 30 minutes
Simply remove the pre-attached gasket (no tools required) and install your own
SterilEnz®-II/EC end caps come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face. SterilEnz®-II/EC end caps are made rugged inert polypropylene suitable for either autoclaving or gamma-irradiation. The caps do not contain any material derived from animal sources. SterilEnz®-II/EC end caps are available in 1"/1.5" style sanitary flange designs.
Pre-attached gaskets are intended for single-use and are discarded with the fitting. Therefore, cleaning validation & potential cross contamination issues are eliminated. SterilEnz®-II/EC Caps work with any standard flange gasket as well. The built-in gasket saves time, money and eliminates gasket preparation by the end-user. An easy hold tab on the fitting allows for easy on/off installation. Our gaskets cannot be misaligned or dropped by operators during assembly. This feature eliminates leaks and potential contamination issues.
USP <88>: Class VI Biological reactivity tests, in vivo.
USP <87>: Cytotoxicity, MEM Elution, in vitro.
USP <661>: Physicochemical Test for Plastics.
USP <1207>: Sterile Product Packaging - Hose Barb Integrity Evaluation, via microbial immersion.
ASTM F756-00 : Rabbit Blood Cell Hemolysis.
SterilEnz®-II/EC are certified ADCF (animal derived component free).