Avantor® Alltima HP C18 EPS, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
- Excellent column stability
- Low to no detectable column bleed
- Multiple selectivity options
Avantor® Alltima HP C18 EPS - Greater retention and enhanced peak symmetry for polar compounds for reversed-phase applications where C18 is too retentive (USP Code: L1).
The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
Customers Who Bought This Also Bought