Avantor® Alltima HP C18 AQ, HPLC Columns
Fournisseur: Avantor
Certificats
A propos de cet article
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
- Excellent column stability
- Low to no detectable column bleed
- Multiple selectivity options
Avantor® Alltima HP C18 AQ - 100% wter wettable for applications requiring high aqueous mobile phases (USP code: L1).
The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
Customers Who Bought This Also Bought