VeriSpec™ Nitrite(N) - NO₂₋ f(N) Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.