VeriSpec™ Nexion KED ICP-MS Calibration Standard for Set Up, 2 Components, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Certified reference material (CRM) for ICP-MS
10 ppb Co, 1 pb Ce
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.