VeriSpec® Multi-Element Standard, 8 Components, RICCA Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Certified reference material (CRM) for ICP-MS
50 ppm K; 10 ppm La, Li, Mn, Na, Sr; 1 ppm Ba, Ca
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.