VeriSpec® Mixed Cation Standard, 6 components, Ricca Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Certified reference material (CRM) for ion chromatography (IC)
50 ppm Li+; 200 ppm Na+, K+, Mg²⁺; 400 ppm NH₄⁺, 1000 ppm Ca²⁺
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.