VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
Certified reference material (CRM) for ion chromatography (IC)
150 ppm SO₄²-, PO₄³; 100 ppm NO₃-, Br-, NO₂-; 30 ppm Cl-, 20 ppm F-
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.