VeriSpec® Strontium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Fournisseur: Avantor
Certificats
A propos de cet article
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.