ContactUsLinkComponent
VeriSpec™ Erbium Standard for ICP 1000 ppm in 2% HNO₃
VeriSpec™ Erbium Standard for ICP 1000 ppm in 2% HNO₃
Supplier:  Avantor
Certificates
VeriSpec™ Erbium Standard for ICP 1000 ppm in 2% HNO₃
VeriSpec™ Erbium Standard for ICP 1000 ppm in 2% HNO₃
Supplier:  Avantor
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.