ContactUsLinkComponent
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl
Fournisseur:  Avantor
Certificats
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl
Fournisseur:  Avantor
Certificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.