VeriSpec® Copper (Cu) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Fournisseur: Avantor
Certificats
A propos de cet article
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.