ContactUsLinkComponent
VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
Fournisseur:  Ricca Chemical
Certificats
VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
Fournisseur:  Ricca Chemical
Certificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.