NORELL™ NMR Tubes, Valved, Chemglass
Fournisseur: Avantor
Certificats
A propos de cet article
NORELL™ Valved NMR Tubes are a completely greaseless PTFE assembly, which is easy to use and to disassemble for cleaning.
- PTFE covered O-ring eliminates material incompatibilities
- Can be handled without flame-sealing tubes
- Includes female joint for quick attachment to vacuum rack
Customers Who Bought This Also Bought