3B Scientific® Breast with Benign Tumor
# de catalogue 470224-932
Fournisseur: American 3B Scientific
Spécifications
- Description:Breast with Tumor
- Height:18 cm
- Cat. no.:470224-932
- Length:25 cm
- Width:18 cm
- Supplier No.:L55/1
- No. of Parts:1
Spécifications
A propos de cet article
For demonstration of ultrasonic B-image mode.
- 3 year manufacturers warranty
Model of a woman’s breast made of 3B SKINlike™ silicone with simulated benign tumor for the demonstration of ultrasonic B-image mode with Ultrasonic Echoscope.
Customers Who Bought This Also Bought