3B Scientific® HIV Viron
# de catalogue 470119-228
Fournisseur: American 3B Scientific
Spécifications
- Description:HIV Viron
- Height:13 cm
- No. of Keyed Items:6
- Cat. no.:470119-228
- Length:19 cm
- Width:13 cm
- Supplier No.:L40
- No. of Parts:2
Spécifications
A propos de cet article
This model of the HI-Virus, enlarged millions of times, shows the outer lipid membrane with protein structures, and the internal nucleus which contains the viral hereditary matter (RNA).
- 3 year manufacturers warranty
The nucleus is removable and condoms can be put underneath to provide a message regarding measures to take in protecting against HIV. No sex education course is complete without talking about HIV, the deadly sexually transmitted disease. The AIDS virus is mounted on base for easy display.
Customers Who Bought This Also Bought