3B Scientific® Prostate Model
# de catalogue 470119-178
Fournisseur: American 3B Scientific
Spécifications
- Description:Prostate
- Height:14 cm
- No. of Keyed Items:29
- Cat. no.:470119-178
- Length:13 cm
- Width:10 cm
- Supplier No.:K41
- No. of Parts:1
Spécifications
A propos de cet article
This prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
- 3 year manufacturers warranty
- Includes 3B Smart Anatomy
The narrowing of the urethra due to the change of the prostate is made clear via the 4 cross sectional views on the bass of the prostate model. The anatomical structures of the prostate model have simple schematic coloring. Prostate model on base.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought