Spécifications
- Description:Rat Brain
- Height:16 cm
- No. of Keyed Items:22
- Cat. no.:470224-890
- Length:14 cm
- Width:10 cm
- Supplier No.:C29
- No. of Parts:2
Spécifications
A propos de cet article
This model shows a rat brain in approx. 6-fold enlargement. Sectioned medially, it can be disassembled into two halves.
- Base includes a diagram of the human brain for comparison
- 3 year manufacturers warranty
The right half of the model shows the structures of the cerebrum, cerebellum and brain stem, each of which is color-coded for didactic purposes (cerebrum = pink, cerebellum = blue, brain stem = yellow), both externally and in the median section.
The left half of the model is largely transparent, thus revealing a view of the colored left lateral ventricle and hippocampus, which can also be seen in the median section. For purposes of comparison, a natural cast of a rat brain and a didactic, small-scale illustration of a human brain in median section are shown on the base, with the same color coding used for the various regions.