3B Scientific® Four Part Brain
# de catalogue 470224-888
Fournisseur: American 3B Scientific
Spécifications
- Description:4 Part Brain
- Height:18 cm
- Cat. no.:470224-888
- Length:14 cm
- Width:14 cm
- Supplier No.:C16
- No. of Parts:5
Spécifications
A propos de cet article
This 4-part brain is medially divided.
- 3 year manufacturers warranty
- Includes 3B Smart Anatomy
All structures of the brain are hand-painted, numbered and identified in a product manual. The brain's right half can be disassembled into: Frontal with parietal lobes , Brain stem with temporal and occipital lobes , Half of cerebellum. This brain is a great educational tool for the human nervous system and anatomy of the brain. This brain is delivered on a removable base.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought