3B Scientific® Muscled Elbow Joint
# de catalogue 470224-876
Fournisseur: American 3B Scientific
Spécifications
- Description:Muscled Elbow Joint
- Height:25 cm
- No. of Keyed Items:45
- Cat. no.:470224-876
- Length:25 cm
- Width:41 cm
- Supplier No.:A883
- No. of Parts:8
Spécifications
A propos de cet article
This Elbow Model with removable muscles by 3B Scientific® is a great tool for student and patient education.
- Muscular origins and insertions are raised and color-coded
- Removable parts: muscle layers can easily be taken off
- Realistic coloring of bone and muscles
- 3 year manufacturers warranty
- Includes 3B Smart Anatomy
Its filigree architecture is determined by influences such as pressure, bending and torsion. Using innovative micro CT technology, we have managed to reconstruct an exact 3-dimensional copy of a piece of cancellous bone from an original and enlarge it 100 times.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought