Muscled Knee Joint
# de catalogue 470224-874
Fournisseur: American 3B Scientific
Spécifications
- Description:Muscled Knee Joint
- Height:33 cm
- No. of Keyed Items:51
- Cat. no.:470224-874
- Length:17 cm
- Width:17 cm
- Supplier No.:A882
- No. of Parts:12
Spécifications
A propos de cet article
The model consists of 12 parts which can be easily disassembled for easy demonstration.
- Muscular origins and insertions are raised and color-coded
- Removable parts: muscle layers can easily be taken off
- Realistic coloring of bone and muscles
- 3 year manufacturers warranty
- Includes 3B Smart Anatomy
The muscles and muscle portions can be taken off, and a special color coding has been used to show muscle origin and insertion points on the femur, tibia, and fibula. Additionally, the model also shows parts of the fibular and tibial collateral ligaments.
Whether your are studying or teaching the deeper anatomical layers of the human knee joint, or are looking for a tool to better educate your patients about their injuries and rehab, the Knee Joint Model by 3B Scientific® is an extraordinary teaching tool and offers exceptional value.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought