ContactUsLinkComponent
3B Scientific® Deluxe Functional Hip Joint
3B Scientific® Deluxe Functional Hip Joint
# de catalogue 470224-868
Fournisseur:  American 3B Scientific
Model Deluxe Functional Hip
3B Scientific® Deluxe Functional Hip Joint
# de catalogue 470224-868
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Deluxe Functional Hip
  • Height:
    32 cm
  • No. of Keyed Items:
    11
  • Cat. no.:
    470224-868
  • Supplier No.:
    A81/1
  • No. of Parts:
    2
Spécifications
A propos de cet article
This high-quality functional hip joint model with ligaments shows the anatomy and possible physiological movements of the human hip joint in exceptional detail.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
This mode clearly demonstrates abduction, anteversion, retroversion, internal and external rotation.
The color of the natural-cast bones is extremely realistic. The cartilage on the hip joint surfaces is marked blue. The functional hip joint consists of portion of the femur and the right half of a pelvis. Mounted on a base for easy display in the classroom or doctors office.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought