ContactUsLinkComponent
3B Scientific® Deluxe Functional Shoulder Joint
3B Scientific® Deluxe Functional Shoulder Joint
# de catalogue 470224-866
Fournisseur:  American 3B Scientific
Model Deluxe Functional Shoulder
3B Scientific® Deluxe Functional Shoulder Joint
# de catalogue 470224-866
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Deluxe Functional Shoulder
  • Height:
    22 cm
  • No. of Keyed Items:
    16
  • Cat. no.:
    470224-866
  • Supplier No.:
    A80/1
  • No. of Parts:
    2
Spécifications
A propos de cet article
This life size deluxe functional shoulder joint model with ligaments shows the anatomy and possible physiological movements of the shoulder joint in exceptional detail.
  • 3 year manufacturers warranty
  • Includes 3B Smart Anatomy
Cleary demonstrate abduction, anteversion, retroversion, internal and external rotation with this high quality shoulder model.
The color of the natural-cast bones in this shoulder model is extremely realistic. The cartilage on the joint surfaces is marked in blue. The shoulder joint model consists of scapula, clavical and a portion of the humerus. Mounted on a base for easy display in the classroom or doctors office.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought