UHPLC Filters, Thermo Scientific
Fournisseur: NATIONAL SCIENTIFIC CO SOV
Certificats
A propos de cet article
These UHPLC filters with low volume filter cartridge design, protect columns for Hypersil GOLD™ 1.9 µm and Syncronis 1.7 µm columns.
- Maintain peak shape
- Provide a low-volume filter cartridge design
- Ensure minimal efficiency loss through dispersion
Customers Who Bought This Also Bought