About this item
The 37 kHz Elmasonic E Plus series includes 9 tank sizes with simple, user-friendly operation. Samples are rapidly mixed, dispersed, dissolved, or homogenized in Pulse mode.
- Thorough and uniform cleaning with Sweep function
- Unique Pulse function performs laboratory mixing and degassing operations
- Pulse function also provides powerful cleaning for difficult jobs
- Quality promise: 2-year warranty with replacment guarantee for the first year
Pulse mode boosts the ultrasonic power to remove tenacious contaminants such as mineral deposits and polishing pastes. Sweep mode produces a continuous slight variation of frequency to provide uniform and thorough cleaning of parts.
The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run.
All models (except the EP10) have adjustable bath temperature from 30°C to 80°C. The control panel indicates when the set temperature has been reached. The timer can be set from 1-30 minutes or for continuous operation with manual shut off. The unit turns off automatically after 8 hours of continuous operation for safety. The 1.5 gallon unit and upwards have a rear drain with a valve on the side of the tank for easy access. The tank cover can be flipped over and used as a drip tray.
Dependable, state-of-the-art Elma ultrasonic cleaners are made in Germany.
Specifications
- Catalog No:
- 75848-748
- 75848-750
- 75848-752
- 75848-754
- 75848-756
- 75848-758
- 75848-740
- 75848-742
- 75848-744
- 75848-746
- Environmentally Preferable:
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Sustainable Packaging, Safer Human and Environmental Health
- Frequency:
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- 37 kHz
- Temperature range:
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- 30°C to 80°C
- Timer:
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- 1-30 minutes or continuous
- Electrical:
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- 115-120V, 60Hz
- Safety:
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation
- Automatic shut down after 8 hours of continuous operation