SPILFYTER® Hazmat Spill Kit Drum, NPS
# de catalogue 15551-794
Fournisseur: NPS
Spécifications
- Description:Hazmat Spill Kit Drum
- Cat. no.:15551-794
- Supplier No.:222800
Spécifications
A propos de cet article
Space-saving option for storing sorbent materials in labs and cleanrooms
- Poly-bagged
- Heat-sealed
- Wall mounted
These wall mount spill kits provide a space-saving option for storing sorbent materials in labs and cleanrooms. All sorbents are poly-bagged and heat-sealed.
Delivery: Kit includes ten 12"x12" Light-Duty Cleanroom Pads, ten 8.5"x17" Medium-Duty Pillows, two Nitrile Gloves, two Poly Aprons, two Splash Goggles, two Disposal Bags and Twist Ties, one Plastic Cabinet w/clear hinged door (Cabinet measures).
Customers Who Bought This Also Bought