Cryokit 400, Cryogenic Gloves, Mapa Professional
Fournisseur: MAPA
Certificats
A propos de cet article
These cryogenic waterproof glove are made of elastic and laminated blue fabric, internal polyester multilayer fleece and polyolefin porelle membrane offer dexterity in very low temperatures.
- Interior finish with knitted thermal protection
- Provide dexterity, comfort and also protection in cryogenic environments
- Polyolefin Porelle membrane keeps hands dry while allowing air to flow
These gloves are suitable for applications handling liquid nitrogen and other cryogenic gases to protect from cold contact and prevent burns from liquid gas leakage.
Customers Who Bought This Also Bought