Coventry™ Sealed Polyester Swabs, ITW Chemtronics®
Fournisseur: ITW Chemtronics
Certificats
A propos de cet article
Swabs feature either knit or nonwoven fabric heads, and are acid and solvent resistant
- High strength, absorbency, solvent compatibility, and absorbency rate
- Does not generate loose fibers or particles
- No adhesives or binders are used in the construction
- Strongest seams available will not split, also good for tight tolerance work
- Micro-fiber has increased number of microspace to trap more particles
Aqua-Prime™ process available on all sealed polyester swabs. Class 10 (ISO Class 4) processing gives low ionic, nonvolatile residue and particle contaminants.
Sealed edges ensure high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes.
Customers Who Bought This Also Bought