Spécifications
- Description:Protective Probe Guard, Plastic, Black
- Supplier No.:081045
Spécifications
A propos de cet article
Provide additional protection and weight with Orion™ Protective Guards for Conductivity Cells and Dissolved Oxygen Probes.
For Use With
Conductivity Cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
Dissolved Oxygen Probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D