Temperature Controller with Process and Limit Control, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Two independent temperature controllers in one cabinet.
- LED is user-selectable for Fahrenheit or Centigrade
Controller has two 900 watt outlets, independent LED display and independent audible over-temp protection alarms. Ideal for saving bench space or for attachment to a support frame through the mounting bracket.
Certifications: cULus rated.
Ordering information: Includes integral rack mounting bracket. One-year Limited Manufacturer’s Warranty.
Spécifications
- Catalog No:
- 75846-504
- 75846-502
- 75846-500
- 75846-498
- 75846-496
- 75846-506
- Accuracy:
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- 0.1% of span, ±1.0 °C, with setpoint ramping feature
- Electrical:
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input
- 120VAC, 15A, 60Hz input