Science Take-Out® Lead An Element Of Danger
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
Follow The Case Of A Young Child With Lead Poisoning
- Available as a single kit with materials for 1 student or group
- Unassembled 10 pack for additional savings
- No additional equipment needed
This complete experiment setup contains all necessary materials and does not require lab equipment. Students will conduct simulated blood tests to determine the child’s blood lead level. They then test samples of dust,
water, soil, and pottery from the child’s home to determine the source of lead. Finally they interpret readings to plan remediation to reduce lead exposure.
Customers Who Bought This Also Bought