Science Take-Out® An Unhealthy Home
Fournisseur: SCIENCE TAKE-OUT, LLC
Certificats
A propos de cet article
Investigate The Source Of A Families Health Problems
- Available as a single kit with materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No additional equipment needed
This complete experiment setup contains all materials and does not require lab equipment. Follow the case of a family suffering from persistent fatigue, respiratory problems, headaches, and mild nausea that began when they moved to an older home. Students perform and analyze simulated strep tests, blood tests, lead tests, and mold tests to determine possible causes for the family’s symptoms.
Customers Who Bought This Also Bought