Red Cabbage Jiffy Juice Extract Powder
# de catalogue 470094-806
Fournisseur: Universe of Science
Numéro CAS:
Spécifications
- Description:Juice Cabbage, 20G Jiffy Juice
- Cat. no.:470094-806
- Supplier No.:2155
Spécifications
A propos de cet article
Red Cabbage Jiffy Juice is a very stable and easy to use red cabbage extract powder. It has low odor and a long shelf-life. This Red Cabbage Jiffy Juice containts red cabbage extract, dextrose, and citric acid and should be kept out of eyes and mouth.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Customers Who Bought This Also Bought