SP Wilmad-LabGlass Powrtwin Temperature Controller, SP Industries
# de catalogue 14212-694
Fournisseur: SP Industries
Spécifications
- Cat. no.:14212-694
- Electrical:120 V, 7A, 840 W (per circuit)
- Supplier No.:LG-8957-100
Spécifications
A propos de cet article
The PowrTwin™ controls two separate circuits independently.
- Output range adjustable from 5-100% of rated voltage
Like the PowrTrol™ (LG-8954-100), the PowrTwin™ is a solid-state, proportional-voltage unit that allows precise manual control of laboratory heating equipment such as heating mantles, tapes and cords.
Ordering information: Includes lighted on-off switch, circuit breaker protection, 3-wire load receptacle and 3-wire line cord with molded plug.
Customers Who Bought This Also Bought