SP Wilmad-LabGlass Minitrol Temperature Controllers, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Regulating apparatus such as heating tapes and mantles that require stepless control, these temperature controllers are manually adjustable.
- Manually adjustable
Each position on the preset dial represents a specific cycle time where the operating voltage load ranges from 5.5 to 100%. Since there is no sensing element, the operator has control over operation. Once desired temperature is determined on initial trial, a simple dial setting controls repeat operations.
Ordering information: Includes 6', 3-wire SJT cord, fuse and pilot light.
Customers Who Bought This Also Bought