Dipping vessel, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Dipping vessel for taking all-layer samples from tanks, tank cars, water courses, water purification plants.
- Equipped with Easy-Flow; a flow optimized flow valve
- High weight (2.1 kg/4.63 lbs.) for faster lowering
- Height with handle is 427 mm (16.81")
- Made of non-sparking brass or stainless steel AISI 304 (1.4301) for Ex area
- Tested for use in the Ex Protection Zone 0
- Explosion Categories IIA, IIB, and IIC
Packaging: Supplied one dipping vessel per package.
Customers Who Bought This Also Bought