Immersion Cylinder, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum, and liquid lubricants in Explosion Categories IIA, IIB, and IIC.
- Screw-off top enables easy cleaning
- High weight for fast lowering
- Robust handle
- Tested for use in the Ex protection zone 0
Completely made from non-spark producing materials, nickel-plated.
The 500 ml model has a weight of 3000 g (105.82 oz.). The 1000 ml model has a weight of 4300 g (15168 oz.).
Packaging: Supplied one immersion cylinder per package.
Customers Who Bought This Also Bought