VACUUBRAND® Service Kit for MZ2C Chemistry-Design Pump, BrandTech
# de catalogue 80086-246
Fournisseur: Brandtech
Spécifications
- Description:Service Kit for VACUUBRAND® MZ2C
- Environmentally Preferable:Sustainable Packaging
- Cat. no.:80086-246
- Supplier No.:696814
Spécifications
A propos de cet article
Set of two diaphragms and four valves.
Does not fit "NT" models.
Customers Who Bought This Also Bought