A propos de cet article
Ideal for laboratory, medical, dental, gas generation, beverage dispensing, and other applications where noise and long life are key requirements.
- Ultra-quiet
- Long-life, two-year-warranty
- Highly efficient
- Easy maintenance
- Integrated filtration
- 100% duty cycle, runs continuously
JUN-AIR has become a leading supplier of oil-less quiet air compressors used in laboratories, dental clinics, medical, and industrial applications worldwide. Our products include a full range of standard and customized compressors and cabinets. Continuous development of products and production facilities helps maintain JUN-AIR’s enviable reputation for quality and reliability among it’s more than half a million satisfied customers.
These compressors comes with few moving parts, which reduces the noise, vibrations as well as wear and tear. The reduced noise and vibration level makes it possible to place the compressor directly at the place of use.