HG Series® Heat Gun Spreader and Reducer, Master Appliance
Fournisseur: Master Appliance
Certificats
A propos de cet article
Professional quality attachments and accessories add versatility to Proheat heat guns.
- Spreader to disperse even heat over a wide surface
- Reducer for targeted heating
Direct, concentrate or reflect heat to allow more versatile and precise applications for heat tool.
Customers Who Bought This Also Bought