LD7 Quarter PMC Pallet Shippers, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
LD7 Quarter PMC Pallet Shippers offer technology, performance, and simplicity.
- Optimize air shipments of temperature-sensitive biologics and pharmaceutical products for durations exceeding five days
- Platform features patent-pending technologies such as our unique interlocking L-shaped panels
- Revolutionary design eliminates corner leaks and drastically reduces edge leaks
- ConvecTECH™ dispersion panels maximize natural convection
- Eliminates side refrigerants, increasing payload and simplifying packing procedures
The new LD7 pallet shipper platform maximizes the space on an LD7 air pallet used in wide-body aircraft. The platform includes solutions for 2 to 8°C, 15 to 25°C and frozen temperature ranges. Each solution has a universal pack-out design to be used in both hot and cold seasons while also accommodating the demands of cross-hemispheric shipments.
BodyTextAccessories: Refrigerant sleeves or dry ice tray are required for each pallet solution configuration. The CRT 15 to 25°C solution requires the pallet shell and 20 units of the refrigerant sleeves requiring ambient conditioning. The refrigerated 2 to 8°C solution requires the pallet shell and 10 units of the refrigerant sleeves requiring refrigerated conditioning and 15 units of the refrigerant sleeves requiring frozen conditioning. The conditioning requirement for each is printed on each sleeve type to simplify the packout procedure. The frozen <-20°C solution requires the pallet shell and the dry ice tray.
Customers Who Bought This Also Bought