Master-Mite® Heat Gun, Master Appliance
# de catalogue 75845-352
Fournisseur: Master Appliance
Spécifications
- Temperature Range:650 °F Max
- Voltage:120V
- Electrical:475W
- Cat. no.:75845-352
- Supplier No.:10008
Spécifications
A propos de cet article
A continuous duty bench-top heating system with 650°F element, stand and heat shrink attachment.
- Quiet running motor, eliminates noisy production areas
- Three optional easy-to-change plug-in heating elements available: 500°, 650° and 800°F
Complete system comes standard with 650°F heating element, adjustable heat gun stand and heat shrink attachment. Ideal heat and air flow (3.8 CFM), performs bench top heating jobs without excess air flow, won’t blow components around work station. Adjustable heat gun is used to activate adhesive, cure epoxies, dry components, melt solder performs, remove decals, shrink tubing and more.
Customers Who Bought This Also Bought