Heating Mantle Micro, Glas-Col®
Fournisseur: Glas-Col
Certificats
A propos de cet article
Precise heat control for small flasks.
- Pliable interior fabric
- Micro size for small samples
- Safe small volume heating
This mantle is used extensively in analytical work and in purification of research products with only small samples available. Fabric exterior provides effective heating in a space saving configuration. Fabric interior softly nestles glass vessels to reduce the chance of thermal shock and damage to glassware. Withstands up to 400 degrees C internal operating temperature.
Delivery: 7-10 days after order.
Customers Who Bought This Also Bought