SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Designed for use with cylindrical reaction kettles LG-8071 through LG-8082, these heating mantles have an aluminum housing exterior.
- Includes detachable 4 foot, 3-wire cord and locking connector
They are equipped with a 3-wire terminal with one grounded to housing.
Customers Who Bought This Also Bought