ContactUsLinkComponent
Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
# de catalogue 75838-694
Fournisseur:  Globe Scientific
Bag Biohazard Specimen Transport 6 in x 10in
Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
# de catalogue 75838-694
Fournisseur:  Globe Scientific
Spécifications
  • Description:
    Biohazard bag, scoreline, and document pouch
  • Size:
    6×9"
  • Cat. no.:
    75838-694
  • Supplier No.:
    4921
Spécifications
A propos de cet article
The special glue closure of our specimen transport bags offer a liquid-tight tamper evident seal.
  • 2 mil gauge polyethylene (PE)
  • Attached document compartment
  • Glue closure tamper evident seal
  • Imprinted with biohazard symbol
  • Perfect for the transport of drug testing products
Simply pull off the liner to expose the glue adhesive and press to close.
Customers Who Bought This Also Bought