Globe Scientific Zipper Closure Specimen Bags, Globe Scientific Inc
Fournisseur: Globe Scientific
Certificats
A propos de cet article
These bags are designed for the safe transportation of specimens and documentation.
- 2 mil gauge polyethylene (PE)
- Attached document compartment
- Zipper closure allows repeated access to specimen
- Imprinted with biohazard symbol
Each bag is printed with the biohazard symbol and offers checkmark boxes to indicate special handling instructions.
Certifications: Complies with OSHA and NCCLS guidelines.
Customers Who Bought This Also Bought