Male Luer-Lok Priming Syringes, Thermo Scientific
Catalogue # 10662-058
Fournisseur: Thermo Scientific
Spécifications
- Volume:1000 µl (1 ml)
- Description:Priming Syringe
- Cat. no.:10662-058
- Supplier No.:365KL531
Spécifications
À propos de cet article
Male Luer-Lok priming syringes offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings