Multi-Voltage Temperature Controller with Audible Alarm and Data Logging, 1200 Watts, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Single outlet, output voltage-selectable temperature controller with audible alarm, and data logging via an RS-485 interface cable (cable sold separately)
- LED is user-selectable for Fahrenheit or Centigrade
Positive stop, multi-voltage dial offers over-voltage protection for Instatherm® products rated at less than 120vac maximum.
Ordering information: Includes integral rack mounting bracket. One-year limited manufacturer's warranty.
Spécifications
- Catalog No:
- 75846-462
- 75846-464
- 75846-466
- 75846-468
- 75846-470
- 75846-472
- Accuracy:
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- Electrical:
- 120V, 10A, 60Hz input, 1200W output
- 120V, 10A, 60Hz input, 1200W output
- 120V, 10A, 60Hz input, 1200W output
- 120V, 10A, 60Hz input, 1200W output
- 120V, 10A, 60Hz input, 1200W output
- 120V, 10A, 60Hz input, 1200W output