Temperature Controller, 1200 Watt, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Single outlet temperature controller with audible alarm
- LED is user-selectable for Fahrenheit or Centigrade
Certifications: cULus safety rated.
Ordering information: Includes integral rack mounting bracket. One-year limited manufacturer's warranty.
Spécifications
- Catalog No:
- 75846-740
- 75846-742
- 75846-744
- 75846-434
- 75846-746
- 75846-738
- Accuracy:
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- 0.1% of span, ± 1.0°C with setpoint ramping feature
- Electrical:
- 120VAC, 10A, 60Hz input, 1200W output
- 120VAC, 10A, 60Hz input, 1200W output
- 120VAC, 10A, 60Hz input, 1200W output
- 120VAC, 10A, 60Hz input, 1200W output
- 120VAC, 10A, 60Hz input, 1200W output
- 120VAC, 10A, 60Hz input, 1200W output